Learn More
Carbosynth LLC. CRF (Ovine) (0.1mg vial), 1ea, MW: 4670.3, Molecular formula: C205H339N59O63S, CAS# 9015-71-8
SDP
Supplier:  Carbosynth LLC. PCR4111S0.1MG
Encompass_Preferred
Synonyms: Ser-Gln-Glu-Pro-Pro-Ile-Ser-Leu-Asp-Leu-Thr-Phe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Met-Thr-Lys-Ala-Asp-Gln-Leu-Ala-Gln-Gln-Ala-His-Ser-Asn-Arg-Lys-Leu-Leu-Asp-Ile-Ala-NH2 Corticotropin-Releasing Factor (Ovine), Storage Temperature: -20°C, References: W. Vale, J. Spiess, C. RivIer, and J. Rivier, Science, 213, 1394 (1981)., C.L. Rivier and P.M. Plotsky, Annu. Rev. Physiol., 48, 475 (1986). (Review), F.A. Antoni, Endocrinol. Rev., 7, 351 (1986). (Review)
Catalog No. 50-168-6398