Learn More
Medchemexpress LLC Cridanimod | 38609-97-1 | MFCD00157050 | 99.8% | 253.26 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW011890100MG
Encompass_Preferred
Cridanimod is a potent progesterone receptor (PR) activator mediated through induction of IFNα and IFNβ expression. It is a small-molecule immunomodulator and interferon inducer. It induces the expression of IFNα and IFNβ, functioning as a small-molecule immunomodulator and interferon inducer.
  • Potent progesterone receptor activator
  • Induces IFNα and IFNβ expression
  • Functions as a small-molecule immunomodulator
  • Interferon inducer
Catalog No. 50-004-55038