Learn More
Evident Crime Scene Products CRIME SCENE RULER 10/PK
SDP
Supplier:  Evident Crime Scene Products 5087W
Encompass
Photo Scale Kit, White, 6 in., Flexible vinyl ruler, Designed for precise, accurate measurements in inches or millimeters, Vinyl rulers have a flat, non-glare finish to eliminate reflections when photographing
Catalog No. NC9249766