Learn More
Chemglass Life Sciences Aluminum Seal, 11mm, Gold, Clear PTFE/Red Rubber
SDP
Supplier:  Chemglass Life Sciences CV3110Y011
Encompass_Preferred
Aluminum Seal, 11mm, Gold, Clear PTFE/Red Rubber. The 11mm anodized aluminum seals are manufactured to fit all 11mm crimp top vials and are available in a variety of colors. Medium durometer red rubber with a .015 layer of clear PTFE. Great and economical for routine analysis. Not recommended for multiple injections.
Catalog No. 50-153-2530