Learn More
World Precision Instrument Crimped Wire Mesh Sterilization Baskets, Tilted Handles-WP-4407CW
SDP
Supplier:  World Precision Instrument WP4407CW
Encompass
Stainless Steel Crimped Wire Mesh Sterilization Baskets with Tilt Handles. Electropolished and burr free to protect wraps and prevent injury. Meets all the requirements of automatic washing and sterilization for all types of instruments.
Catalog No. 50-209-0257