Learn More
Medchemexpress LLC Crinecerfont hydroch 25mg | 321839-75-2 | 25MG
SDP
Supplier:  Medchemexpress LLC HY106203A25MG
Encompass_Preferred
Crinecerfont (SSR-125543) hydrochloride is a potent orally active non-peptide CRF1 receptor antagonist Crinecerfont can be used for Classic congenital adrenal hyperplasia (CAH) research[1 Crinecerfont (hydrochloride) is a click chemistry reagent it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-002-43526