Learn More
Medchemexpress LLC cRIPGBM chloride 10mM 1mL | 2361988-77-2 | 446.90 | C26H20ClFN2O2 | 1 ML

Supplier: Medchemexpress LLC HY11563010MM 1ML
cRIPGBM chloride is a small-molecule RIPK2 modulator supplied as a 10 mM solution in DMSO for research use. The product is provided as a 1 mL solution intended for in vitro assays and screening.
- Supplied as a 10 mM solution in DMSO
- Provided as a 1 mL solution
- High purity (99.82%)
- Molecular formula C26H20ClFN2O2 and molecular weight 446.90 g·mol⁻¹
- CAS number 2361988-77-2
- For research use only; not for human or clinical applications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.