Learn More
Sino Biological Recombinant Streptococcus pyogenes CRISPR-Cas9 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 40572A08B200
Encompass_Preferred
A DNA sequence encoding the s. pyogenes serotype M1 cas9 (WP_010922251.1) (Met1-Asp1368) was expressed with a nuclear localization sequence (NLS) at the N-terminus, two NLSs and a polyhistidine tag at the C-terminus.
Catalog No. NC1727187