Learn More
Medchemexpress LLC HY-50878 100mg , Crizotinib CAS:877399-52-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50878100MG
Encompass_Preferred
HY-50878 100mg Crizotinib CAS: 877399-52-5 Crizotinib (PF-02341066) is an orally bioavailable, ATP-competitive ALK and c-Met inhibitor with IC50s of 20 and 8 nM, respectively. Crizotinib inhibits tyrosine phosphorylation of NPM-ALK and tyrosine phosphorylation of c-Met with IC50s of 24 and 11 nM in cell-based assays, respectively. Crizotinib is also a ROS1 inhibitor. Crizotinib has effective tumor growth inhibition. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9565