Learn More
Selleck Chemical LLC Crizotinib S1068-200mg
SDP
Supplier:  Selleck Chemical LLC S1068200MG
Encompass_Preferred
Crizotinib is a potent inhibitor of c-Met and ALK with IC50 of 11 nM and 24 nM in cell-based assays respectively It is also a potent ROS1 inhibitor with Ki value less than 0 025 nM Crizotinib induces autophagy through inhibition of the STAT3 pathway in multiple lung cancer cell lines
Catalog No. 50-310-8199