Learn More
TARGETMOL CHEMICALS INC Crizotinib 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1661500MG
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Crizotinib (PF-02341066) is an ATP-competitive small-molecule tyrosine kinase inhibitor of c-MET (IC50 8 nM) and ALK (IC50 20 nM) receptors. Purity 99.35%
Catalog No. 50-287-7395