Learn More
Medchemexpress LLC Crizotinib | 877399-52-5 | 99.7% | C21H22Cl2FN5O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY508785MG
Encompass_Preferred
Crizotinib is an orally bioavailable, ATP-competitive ALK, c-Met, and ROS1 inhibitor. It inhibits tyrosine phosphorylation of NPM-ALK and c-Met in cell-based assays. This compound demonstrates effective tumor growth inhibition and is for research use only, not for sale to patients.
  • Orally bioavailable and ATP-competitive.
  • Inhibits ALK, c-Met, and ROS1.
  • Demonstrates effective tumor growth inhibition.
  • Inhibits tyrosine phosphorylation of NPM-ALK and c-Met.
  • Purity of 99.7%.
  • Stable under recommended storage conditions.
  • Suitable for research applications.
Catalog No. 50-003-58172