Learn More
Selleck Chemical LLC Crizotinib hydrochloride S5190-25mg
SDP
Supplier:  Selleck Chemical LLC S519025MG
Encompass_Preferred
Crizotinib (PF-02341066) hydrochloride (Xalkori) inhibits tyrosine phosphorylation of c-Met and nucleophosmin (NPM)-anaplastic lymphoma kinase (ALK) with IC50 of of 11 nM and 24 nM in cell-based assays respectively Crizotinib hydrochloride is also a potent ROS1 inhibitor with Ki less than 0 025 nM Crizotinib induces autophagy through inhibition of the STAT3 pathway in multiple lung cancer cell lines
Catalog No. 50-311-2255