Learn More
ABclonal Technology CRKL Rabbit pAb
SDP
Supplier:  ABclonal Technology A117351000UL
Encompass_Preferred
This gene encodes a protein kinase containing SH2 and SH3 (src homology) domains which has been shown to activate the RAS and JUN kinase signaling pathways and transform fibroblasts in a RAS-dependent fashion. It is a substrate of the BCR-ABL tyrosine kinase, plays a role in fibroblast transformation by BCR-ABL, and may be oncogenic.
Catalog No. 50-001-16109