Learn More
Medchemexpress LLC Crocin | 42553-65-1 | 976.96 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0697R
Encompass_Preferred
Crocin (Standard) is an analytical standard of Crocin, intended for research and analytical applications. Crocin (Crocin I) is an orally active natural product isolated from the stigma of Crocus sativus. It demonstrates various biological activities, including the inhibition of tumor cell proliferation, promotion of apoptosis through the JAK pathway, and anti-inflammatory, antioxidant, and antitumor properties.
  • Analytical standard for research and analytical applications
  • Natural product isolated from Crocus sativus
  • Inhibits tumor cell proliferation and promotes apoptosis
  • Exhibits anti-inflammatory, antioxidant, and antitumor activities
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Suitable for applications such as HPLC, GC, and MS
Catalog No. 50-003-90215