Learn More
Sigma Aldrich Fine Chemicals Biosciences Crocin phyproofR Reference10MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences PHL8039110MG
Encompass
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG
Catalog No. 50-290-0445