Learn More
Medchemexpress LLC HY-13603 5mg Medchemexpress, Crolibulin CAS:1000852-17-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY136035MG
Encompass_Preferred
Medchemexpress, HY-13603 5mg Crolibulin CAS:1000852-17-4 Crolibulin (EPC2407) is a tubulin polymerization inhibitor, with potent apoptosis induction and cell growth inhibition. Crolibulin has anti-tumor activity. Crolibulin also has cardiovascular toxicity and neurotoxicity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1418