Learn More
Cayman Chemical -CromakalIm 10mg
SDP
Supplier:  Cayman Chemical 1460110 MG
Encompass_Preferred
An activator of the ATP-sensitive K+ channels that displays anti-ischemic (EC25 = 8.9 μM) and cardioprotective (IC50 = 0.03 μM) effects in rat hearts
Catalog No. 50-296-9679