Learn More
Medchemexpress LLC Cromakalim | 94470-67-4 | 99.4% | 286.33 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY110011100MG
Encompass_Preferred
Cromakalim is a potassium channel opener that can be utilized as a bronchodilator for asthma. It inhibits the spontaneous tone of human isolated bronchi in a concentration-dependent manner, demonstrating an effectiveness comparable to isoprenaline or theophylline.
  • Potassium channel opener
  • Can be used as a bronchodilator in asthma
  • Inhibits the spontaneous tone of human isolated bronchi
Catalog No. 50-003-46388