Learn More
Cayman Chemical Crotalara palda Usaramn 10mg
SDP
Supplier:  Cayman Chemical 3489410 MG
Encompass_Preferred
A pyrrolizidine alkaloid with antibacterial and hepatotoxic activities; reduces S. epidermidis biofilm formation at 1 mg/ml; induces liver necrosis in rats at 0.05, 0.1, and 0.2 mmol/kg
Catalog No. 50-291-1333