Learn More
Enterprise Technology Solutions Crown Mats & Matting Wiper Mat, Oxford, 36"x60", Black/Gray
SDP
Supplier:  Enterprise Technology Solutions CWNOXH035GY
Encompass_Preferred
Beautifully designed, this mat combines performance and appearance for reliable performance that looks great in any setting. Heavyweight, loop-pile design effectively keeps floors clean from dirt and debris. Attractive pattern features a combination of abrasive and sturdy olefin fibers for well-balanced wiping and scraping performance. Mat is designed for use in medium-traffic to heavy-traffic areas.
Catalog No. 50-008-31483