Learn More
Genscript Corporation CRP 2R4 mAb Mouse
Supplier:  Genscript Corporation V0150510
Encompass
C-reactive protein (CRP) is synthesized in the liver by macrophage Its level in blood increases when inflammation occurs CRP serves as a useful marker for diagnosis of inflammation High-sensitivity C-reactive protein (hsCRP) can serve as a risk marker for the diagnosis of myocardial infarction ischemic stroke peripheral vascular disease Measuring hsCRP in blood is helpful to provide positive treatment of the disease
Catalog No. 50-000-57339