Learn More
Bachem C-Reactive Protein (CRP) (174-185)
SDP
Supplier:  Bachem 4018347.0005
Encompass_Preferred
C-Reactive Protein (CRP) (174-185), H-Ile-Tyr-Leu-Gly-Gly-Pro-Phe-Ser-Pro-Asn-Val-Leu-OH, 147516-85-6, C62H93N13O16, Mr 1276.50, 5mg. RS-83277. CRP, an acute phase serum protein synthesized in response to inflammatory cytokines, produces antitumor effects in animals. The fragment CRP (174-185) (RS-83277) shows similar activity by significantly enhancing the tumoricidal activity of human monocytes and alveolar macrophages in vitro. RS83277 may therefore play a role as a biological response modifier in cancer therapy. Former CAS-number 160369-86-8.
Catalog No. 50-194-6702