Learn More
SEKISUI DIAGNOSTICS LLC CPR Ultra Wide Range Reagent Kit | 1x30 mL (R1); 1x20 mL (R2)
Supplier:  SEKISUI DIAGNOSTICS LLC 082
Encompass_Preferred
C-reactive protein is described as an acute phase protein that is involved in the activation of complement, acceleration of phagocytosis, and detoxification of substances released from damaged tissue. CRP is one of the most sensitive indicators of inflammation. In response to an inflammatory stimulus, a rise in CRP may be detected within 6 hours. CRP is a sensitive, non-specific indicator of acute phase reactants. The level of CRP in serum is elevated in patients with arthritis or liver disease such as hepatitis A, hepatitis B, or biliary cirrhosis, and after severe infections such as septic shock. The CRP Ultra Wide Range Reagent kit is an immunoassay for the quantitative measurement of C-reactive protein in serum and plasma samples. Measurement of C-reactive protein is useful in the detection and evaluation of infection, tissue injury and inflammatory disorders. Wide linear range: 0.05 mg/L to 160 mg/L. Accurate: Calibrators standardized against CRM 470.
Catalog No. NC1595039