Learn More
Trajan Scientific CRS ONE o-rings for SGE inlet liners, FKM, P005 PK10
SDP
Supplier:  Trajan Scientific 0726783
Encompass_Preferred
CRS ONE o-rings for SGE inlet liners, FKM, P005 PK10
Catalog No. 50-272-5023