Learn More
Medchemexpress LLC CRT (Iron Peptide Mimic) | 2414254-70-7 | 99.8% | 933.11 | 10 MG

Supplier: Medchemexpress LLC HYP553310MG
CRT is an iron peptide mimic that binds to apo-transferrin (apo-Tf). It can be used to modify nanoparticles and enhances drug delivery efficiency.
- Solid appearance
- White to off-white color
- Sequence: Cys-Arg-Thr-Ile-Gly-Pro-Ser-Val-Cys (Cys1-Cys5)
- Formula: C37H64N12O12S2
- Soluble in DMSO at 14.29 mg/mL (15.31 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.