Learn More
Medchemexpress LLC CRT0063465 | C17H16BrN3O2 | 5 MG

Supplier: Medchemexpress LLC HY1539175MG
CRT0063465 is a ligand of Human PGK1 and Stress Sensor DJ1, with a Kd of 24 μM of PGK1. It modulates the shelterin complex composition and telomere length.
- Purity: 99.64%
- Molecular weight: 374.23
- Appearance: Solid
- Color: Off-white to light yellow
- Target: Phosphatase
- Pathway: Metabolic Enzyme/Protease
- Solubility (in vitro): DMSO: 20 mg/mL (53.44 mM)
- Storage (solid): 4°C, protect from light
- Storage (in solvent): -80°C for 6 months; -20°C for 1 month (protect from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.