Learn More
TARGETMOL CHEMICALS INC CRT0066101 DIHYDROCHLORI 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TQ010410MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CRT0066101 dihydrochloride is an effective and selective PKD inhibitor (IC50s 1 2.5 and 2 nM for PKD1 2 and 3). purity: 99%
Catalog No. 50-257-6159