Learn More
Medchemexpress LLC CRT0066101 dihydrochloride | 1883545-60-5 | 99.9% | 411.33 g/mol | C18H24Cl2N6O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15698A10MG
Encompass_Preferred
CRT0066101 dihydrochloride is a small-molecule dihydrochloride salt reported as a potent and orally active protein kinase D (PKD) inhibitor for use in biochemical and cellular research. It is provided at high reported purity and available in multiple solid and solution formats to support laboratory experiments.
  • Potent PKD inhibition with low-nanomolar IC50 values for PKD1-3.
  • Supplied as a dihydrochloride salt for improved solubility in aqueous and DMSO solutions.
  • High purity suitable for biochemical assays and cellular studies.
  • Multiple pack sizes and solution formats for flexible experimental needs.
  • Recommended storage: sealed, away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-88266