Learn More
Selleck Chemical LLC CRT0066101 dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S836625MG
Encompass_Preferred
CRT0066101 dihydrochloride is a small molecule PKD family specific inhibitor which specifically blocks PKD1/2 activity and does not suppress PKC /PKC /PKC activity in multiple
Catalog No. 50-000-01096