Learn More
Selleck Chemical LLC CRT0066101 dihydrochloride 5mg
SDP
Supplier:  Selleck Chemical LLC S83665MG
Encompass_Preferred
CRT0066101 dihydrochloride is a small molecule PKD family specific inhibitor which specifically blocks PKD1/2 activity and does not suppress PKC?/PKC?/PKC? activity in multiple. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3355