Learn More
Abcam CRT0066101 (mM/ml), protein kinase D inhibitor, 20MM_ML
Supplier:  Abcam AB14674020MM_ML
Encompass_Preferred
MW 411.33 g/mol, Purity >99%. Potent and selective protein kinase D inhibitor (IC₅₀ values are 1, 2 and 2.5 nM at PKD1, PKD3 and PKD2, respectively). Reduces proliferation of pancreatic cancer cells. Orally active. Antitumor agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4503