Learn More
Abcam CRT0066101, protein kinase D inhibitor, 10MG
Supplier:  Abcam AB14463710MG
Encompass_Preferred
MW 411.3 Da, Purity >99%. Potent and selective protein kinase D inhibitor (IC₅₀ values are 1, 2 and 2.5 nM at PKD1, PKD3 and PKD2, respectively). Reduces proliferation of pancreatic cancer cells. Orally active. Antitumor agent. Active in vivo and in vitro. Also available in simple stock solutions (ab146740) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5543