Learn More
TARGETMOL CHEMICALS INC CRT0066854 HYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T108885MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CRT0066854 hydrochloride is an effective selective atypical PKCs inhibitor. CRT0066854 is against full-length (FL) PKC(iota) PKC(zeta) and ROCK-II kinases with IC50 values of 132 nM 639 nM and 620 nM respectively. purity: 97%
Catalog No. 50-258-1627