Learn More
Novus Biologicals 66-440 kDa Separation Module, 8 x 13 capillary cartridges
SDP
Supplier:  Novus Biologicals SMW006
Encompass
Compatible with Jess, Abby and Wes systems. Includes separation box with Capillary Cartridges, 66-440 kDa Pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer, plus Standard Pack 1 clamshell with DTT, FL Standard master mix with 29 kDa System Control protein. Compatible with Detection module - sold separately. Sizing: 8x13 capillaries per cartridge (104 data points).
Catalog No. NC1219861