Learn More
Ellsworth Adhesive Co PPG Semco 229734 Cartridge without Plunger HD 20 oz
Supplier:  Ellsworth Adhesive Co 229734
Encompass
PPG Semco 229734 Cartridge without Plunger HD is a disposable, high density, polyethylene cartridge that is used for storage and application of chemical fluids such as adhesives, epoxies, urethanes, acrylics, solder paste, or polysulfides. It can be used with pneumatic and manual dispensing systems. All cartridges have a 1/4 in NPT female thread dispensing orifice. Semco components are recommended for use with other Semco parts only. 20 oz.
Catalog No. NC3242919