Learn More
Med Vet International Cruciate Repair Crimping Forceps, 7.5in, J0710
Supplier:  Med Vet International JOR710
Encompass_Preferred
Designed with a flattened area on the forcep jaws that crimps the metal tubes evenly without risk of cutting the nylon suture.
Catalog No. 50-005-84600