Learn More
Hach Company Crucible Holder, Walter
Supplier:  Hach Company 2233200
Encompass
Rubber with glass stem ground at 45° angle. Holds 25 mL Gooch Crucible. Fits filter flasks that take stopper sizes 6-8.
Catalog No. 50-199-5589