Learn More
Medchemexpress LLC Crustecdysone (20-Hydroxyecdysone) | 5289-74-7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN6979100MG
Encompass_Preferred
Crustecdysone (20-Hydroxyecdysone) is a naturally occurring ecdysteroid hormone isolated from Serratula coronata. It controls the ecdysis (moulting) and metamorphosis of arthropods, inhibits caspase activity, and induces autophagy via the 20E nuclear receptor complex, EcR-USP. It also exhibits regulatory or protective roles in the cardiovascular system and is an active metabolite of Ecdysone.
  • Controls ecdysis (moulting) and metamorphosis of arthropods
  • Inhibits caspase activity
  • Induces autophagy via the 20E nuclear receptor complex, EcR-USP
  • Exhibits regulatory or protective roles in the cardiovascular system
  • Active metabolite of ecdysone
  • Induces cell autophagy and caspase-3 activity in Drosophila fat body
  • Reduces blood pressure and prevents dilated cardiac hypertrophy in SHR rats
Catalog No. 50-004-19849