Learn More
Medchemexpress LLC Crustecdysone (20-Hydroxyecdysone) | 5289-74-7 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN697950MG
Encompass_Preferred
Crustecdysone (20-Hydroxyecdysone) is a naturally occurring ecdysteroid hormone isolated from Serratula coronata. It controls the ecdysis (moulting) and metamorphosis of arthropods, inhibits caspase activity, and induces autophagy via the 20E nuclear receptor complex, EcR-USP. It also exhibits regulatory or protective roles in the cardiovascular system. Crustecdysone is an active metabolite of ecdysone.
  • Naturally occurring ecdysteroid hormone
  • Isolated from Serratula coronata
  • Controls ecdysis (moulting) and metamorphosis of arthropods
  • Inhibits caspase activity
  • Induces autophagy via the 20E nuclear receptor complex, EcR-USP
  • Exhibits regulatory or protective roles in the cardiovascular system
  • Active metabolite of ecdysone
  • High purity (99.20%)
Catalog No. 50-004-17950