Learn More
Thomas Scientific Cryo Laser Labels, 1.26" x 0.87", Removable, White
SDP
Supplier:  Thomas Scientific 1189F70
Encompass
Cryogenic plastic labels for long-term storage in liquid nitrogen and deep-freezing harsh environments for biomedical research and clinical laboratories. Cryo labels accept desktop laser printing and permanent marker. Water-resistant; Wear and tear proof; Do not jam printers; Resistant to UV and Gamma irradiation.
Catalog No. NC0244076