Learn More
GA International Dual Point Cryogenic Waterproof Permanent Marker Brown 6/pack
Supplier:  GA International MP1BR6
Encompass_Preferred
Waterproof, cryogenic permanent marker with dual point, fine and extra-fine high-quality tips. This 2-in-1 marker is available in a range of colors, and has excellent performance on plastic, glass, metal, and cloth surfaces. Ideal for use in bio-medical laboratories, this permanent marker can be used to write on cryogenic labels, for subsequent storage in liquid nitrogen, and lab freezers.
Catalog No. NC2394830