Learn More
Research Products International Corp NitroTag CryoCombo Barcode Labels, 1 Inch Core, Yellow, 2000 Labels per Roll
SDP
Supplier:  Research Products International Corp 247189Y
Encompass
NitroTag labels with superior permanent adhesive adhere to wet surfaces and can immediately be submerged into liquid nitrogen. Recommended for long-term storage and biopreservation of cryogenic vials in liquid and vapor phase nitrogen; liquid helium; -80° freezer and dry ice transportation. Labels accept all forms of thermal transfer printing and are alcohol resistant.
Catalog No. 50-153-7684