Learn More
GA International Cryogenic Barcode Labels for Liquid Nitrogen Storage with SimPEEL™ Technology (Patent Pending) 1" x 1" / 25.4mm x 25.4mm Rectangle White From -196°C to +110°C (-321°F to +230°F)
Supplier:  GA International SPJTTA29NPC11WH
Encompass_Preferred
Cryogenic labels for identifying small diameter cryo vials, tubes, plates, cardboard freezer boxes, polypropylene cryo-boxes, and plastic bags. These cryo labels feature our new patent-pending SimPEEL™ technology, making peeling them off their liner quick and easy. Their innovative adhesive-free tab design also makes the label’s application on their destined container hassle-free, as they won’t stick to your hands or gloves, eliminating in this fashion adhesive contamination. Ideal for routine, high-volume container identification in hospitals, clinics, blood collection centers, biotechnology companies, biorepositories, and others. They are designed for long-term cryo storage under cryogenic laboratory conditions, including liquid nitrogen (-196°C/-321°F), freezers (-80°C/-112°F), and for dry ice transport. Labels can be printed with all major brands of thermal-transfer printers.
Catalog No. NC2740881