Learn More
GA International Cryogenic Barcode Labels – 1″ x 0.4375″ #JTTA-1 Core size: 1", Pink, QTY: 1000
Supplier:  GA International JTTA1C11PI
Encompass_Preferred
NitroTAG® – Cryo Barcode Labels for Liquid Nitrogen StorageResistant to long-term cryogenic storage in freezers and liquid nitrogen (-196°C/-321°F)Perfect for cryovials, tubes, cryo-boxes, freezer-boxes, and many other cryo-containersResistant to spraying and swabbing with alcohol and surface sanitizing wipesRequire no cure time before liquid nitrogen immersion. Will also adhere to wet surfaces.Ideal for serialized data and barcodes, thanks to high-quality thermal-transfer printingAvailable in a large range of sizes, colors, and configurations.
Catalog No. NC2749570