Learn More
Med Vet International Jorvet Cryogun Attachments, 18mm Cryochamber, J0810DD
Supplier:  Med Vet International JOR810DD
Encompass_Preferred
Permits precise confined spray. Liquid nitrogen pools over deep lesions. Suggested use with malleable extensions (e.g., JOR810ra)
Catalog No. 50-004-95865