Learn More
Med Vet International Jorvet Cryogun Attachments, 2mm Conical-Shaped Closed Probe, J0810EB
Supplier:  Med Vet International JOR810EB
Encompass_Preferred
These probes are inserted into large or ulcerated lesions, thereby markedly increasing the surface area of the freeze. Closed Probe. Only the tip is cooled via a hollow tube down the probe. A side port vents the liquid nitrogen.
Catalog No. 50-004-95836