Learn More
Mitegen Autogrid Tweezers From Nanosoft
SDP
Supplier:  Mitegen MCEMNSAGTW01
Encompass
Autogrid Tweezers from Nanosoft are engineered for seamless integration with autogrid assembly and cassette loading stations used in Thermo Fisher Scientific (TFS) autoloader-based cryo-EM workflows. These tweezers feature the same precise geometry as the TFS version, ensuring that cartridges and autogrids are handled and loaded into cassettes identically. The NanoSoft Autogrid Tweezers are an essential tool for efficient and precise cryo-EM sample handling and preparation, specifically compatible with Thermo Fisher Scientific autoloader systems.
Catalog No. NC1863627