Learn More
Andwin Scientific CRYO GLOVES MIDARM MD EA
SDP
Supplier:  Andwin Scientific 71074
Encompass_Preferred
The medium Mid Arm Cryo-Gloves, are ideal for use in clinical laboratory, bio-medical, blood banks, frozen food processing, and pharmaceutical environments. The gloves can protect users from 135C to 150C. They are water-resistant and are comfortable to wear with its lightweight design.
Catalog No. 50-727-521