Learn More
Alkali Scientific ASI™ Cryo Labels Rolls, 13mm Dots for 1.5-2mL Tubes, Orange, 1000, 1 Roll/Cs
SDP
Supplier:  Alkali Scientific LCD12O
Encompass
We offer cryogenic storage Cryo Labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).  
Catalog No. 50-302-7036